1,2,3-Trimethoxybenzene 98%

Supplier: Ambeed
Warning

A130157-100G A130157-10G A130157-25G A130157-500G A130157-5G
77190-020EA 20 USD
77190-020 77190-022 77190-024 77190-026 77190-028
1,2,3-Trimethoxybenzene 98%
1,2,3-Trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MDL Number: MFCD00008358
CAS Number: 634-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR