2-Bromo-5H-pyrrolo[2,3-b]pyrazine-7-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A130133-100MG
A130133-1G
A130133-250MG
77161-844EA
43.26
USD
77161-844
77161-846
77161-848
2-Bromo-5H-pyrrolo[2,3-b]pyrazine-7-carbaldehyde 95%
2-Bromo-5H-pyrrolo[2,3-b]pyrazine-7-carbaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrN₃O MW: 226.03 g/mol |
MDL Number:
MFCD17015976 CAS Number: 1185428-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...