3-Amino-1-benzylpiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylpiperidin-3-amine, 1-Benzylpiperidin-3-ylamine

A129898-1G A129898-250MG
77161-836EA 18.54 USD
77161-836 77161-838
3-Amino-1-benzylpiperidine 98%
3-Amino-1-benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD04038444
CAS Number: 60407-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR