3-Bromo-1H-pyrrolo[2,3-c]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-azaindole

A129458-1G A129458-250MG A129458-25G A129458-5G A129458-10G
77161-812EA 36.05 USD
77161-812 77161-814 77161-816 77161-818 77683-149
3-Bromo-1H-pyrrolo[2,3-c]pyridine 97%
3-Bromo-1H-pyrrolo[2,3-c]pyridine

Precaución: For research use only.

Formula: C7H5BrN2
MW: 197.0300 g/mol
MDL Number: MFCD06738910
CAS Number: 67058-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR