Methyl 2-amino-6-chlorobenzoate hydrochloride 95%

Supplier: Ambeed
Warning

A129278-1G A129278-250MG A129278-5G
77161-806EA 9 USD
77161-806 77161-808 77161-810
Methyl 2-amino-6-chlorobenzoate hydrochloride 95%
Methyl 2-amino-6-chlorobenzoate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉Cl₂NO₂
MDL Number: MFCD28101674
CAS Number: 1619264-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR