5-Hydroxy-1H-indole-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A129011-250MG A129011-5G A129011-1G
77161-794EA 70.04 USD
77161-794 77161-796 77683-144
5-Hydroxy-1H-indole-3-carbaldehyde 97%
5-Hydroxy-1H-indole-3-carbaldehyde

Precaución: For research use only.

Formula: C₉H₇NO₂
MW: 161.157 g/mol
MDL Number: MFCD06804528
CAS Number: 3414-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR