2-Amino-3-(3,4,5-trifluorophenyl)propanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A128988-1G A128988-250MG
77190-004EA 32 USD
77190-004 77190-006
2-Amino-3-(3,4,5-trifluorophenyl)propanoic acid hydrochloride 97%
2-Amino-3-(3,4,5-trifluorophenyl)propanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₉ClF₃NO₂
MDL Number: MFCD29038823
CAS Number: 870483-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR