(5-Methylpyridin-2-yl)methanamine 97%

Supplier: Ambeed
Danger

A128932-100MG A128932-10G A128932-1G A128932-250MG A128932-25G A128932-5G
77161-782EA 11 USD
77161-782 77161-784 77161-786 77161-788 77891-906 77891-907
(5-Methylpyridin-2-yl)methanamine 97%
(5-Methylpyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MDL Number: MFCD08752541
CAS Number: 45715-08-0
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR