Sodium pivalate 95%

Supplier: Ambeed
Warning

A128911-100G A128911-10G A128911-1G A128911-250MG A128911-25G A128911-5G
77161-770EA 237.01 USD
77161-770 77161-772 77161-774 77161-776 77161-778 77161-780
Sodium pivalate 95%
Sodium pivalate

Precaución: for Research Use Only

Formula: C₅H₉O₂·Na
MDL Number: MFCD00064202
CAS Number: 1184-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR