3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one acetate 97%

Supplier: Ambeed
Warning

A128735-100MG A128735-1G A128735-250MG
77161-750EA 117.01 USD
77161-750 77161-752 77161-754
3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one acetate 97%
3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O·CH₃CO₂H
MDL Number: MFCD25541770
CAS Number: 1346575-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR