Methyl 3-bromo-5-(trifluoromethyl)picolinate 97%

Supplier: Ambeed
Warning

A128636-100MG A128636-1G A128636-250MG
77189-992EA 34 USD
77189-992 77189-994 77189-996
Methyl 3-bromo-5-(trifluoromethyl)picolinate 97%
Methyl 3-bromo-5-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₅BrF₃NO₂
MW: 284.03 g/mol
MDL Number: MFCD14698000
CAS Number: 1211536-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR