2-Methylpiperidine-3-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A128529-100MG A128529-1G A128529-250MG
77161-744EA 64.89 USD
77161-744 77161-746 77161-748
2-Methylpiperidine-3-carboxylic acid hydrochloride 95%
2-Methylpiperidine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MW: 179.65 g/mol
MDL Number: MFCD15474934
CAS Number: 1220040-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR