3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-6-(methylamino)pyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A128495-1G A128495-250MG A128495-5G
77161-738EA 77.25 USD
77161-738 77161-740 77161-742
3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-6-(methylamino)pyrimidine-2,4(1H,3H)-dione 97%
3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-6-(methylamino)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₃FIN₃O₂
MW: 401.17 g/mol
MDL Number: MFCD18207184
CAS Number: 871700-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR