4-(4-Methylpyrazol-1-yl)benzylamine 97%

Supplier: Ambeed
Warning

Synonyms: (4-(4-Methyl-1H-pyrazol-1-yl)phenyl)methanamine

A128418-1G A128418-250MG
77189-978EA 272.95 USD
77189-978 77189-980
4-(4-Methylpyrazol-1-yl)benzylamine 97%
4-(4-Methylpyrazol-1-yl)benzylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃
MW: 187.25 g/mol
MDL Number: MFCD08699675
CAS Number: 1006951-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR