1,3,5-Tris(3-bromophenyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 3,3''-Dibromo-5'-(3-bromophenyl)-1,1':3',1''-terphenyl

A128386-1G A128386-250MG A128386-25G A128386-5G
77189-970EA 12 USD
77189-970 77189-972 77189-974 77189-976
1,3,5-Tris(3-bromophenyl)benzene 97%
1,3,5-Tris(3-bromophenyl)benzene

Precaución: for Research Use Only

Formula: C₂₄H₁₅Br₃
MDL Number: MFCD02323390
CAS Number: 96761-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR