3-Amino-6-(4-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-N-(pyridin-3-yl)pyrazine-2-carboxamide 98+%

Supplier: Ambeed
Warning

A128237-10MG A128237-1MG A128237-5MG
77161-730EA 106 USD
77161-730 77161-732 77161-734
3-Amino-6-(4-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-N-(pyridin-3-yl)pyrazine-2-carboxamide 98+%
3-Amino-6-(4-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-N-(pyridin-3-yl)pyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₁H₂₃N₇O₃S
MDL Number: MFCD26397072
CAS Number: 486424-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR