8-Isoquinolinecarboxylic acid 95%

Supplier: Ambeed
Warning

A128028-100MG A128028-10G A128028-1G A128028-250MG A128028-5G A128028-25G
77161-708EA 13 USD
77161-708 77161-710 77161-712 77161-714 77161-716 77891-899
8-Isoquinolinecarboxylic acid 95%
8-Isoquinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MDL Number: MFCD06738935
CAS Number: 61563-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR