3,5,6-Trichloropyrazine-2-carbonitrile 95%

Supplier: Ambeed
Warning

A127983-100MG A127983-250MG
77161-696EA 112.27 USD
77161-696 77161-698
3,5,6-Trichloropyrazine-2-carbonitrile 95%
3,5,6-Trichloropyrazine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅Cl₃N₃
MW: 208.43 g/mol
MDL Number: MFCD28099394
CAS Number: 30818-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR