5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Diamino-1-methyluracil

A127924-1G A127924-250MG A127924-5G A127924-100MG
77161-690EA 79 USD
77161-690 77161-692 77161-694 77891-898
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione 95%
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₅H₈N₄O₂
MW: 156.15 g/mol
MDL Number: MFCD00091924
CAS Number: 6972-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR