4,5-Dihydro-6-methyl-3(2H)-pyridazinone 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-4,5-dihydropyridazin-3(2H)-one

A127809-100G A127809-10G A127809-1G A127809-25G A127809-5G
77161-680EA 84.46 USD
77161-680 77161-682 77161-684 77161-686 77161-688
4,5-Dihydro-6-methyl-3(2H)-pyridazinone 97%
4,5-Dihydro-6-methyl-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₅H₈N₂O
MW: 112.13 g/mol
MDL Number: MFCD00006455
CAS Number: 5157-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR