(S)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A127745-10G A127745-25G A127745-5G A127745-1G
77161-674EA 42.23 USD
77161-674 77161-676 77161-678 77683-132
(S)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride 97%
(S)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride

Precaución: For research use only.

Formula: C₆H₁₂ClNO₂
MW: 165.61 g/mol
MDL Number: MFCD04115802
CAS Number: 1508261-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR