2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: Mirtazapine carboxylic acid

A127489-100MG A127489-1G A127489-250MG A127489-5G
77189-938EA 22.66 USD
77189-938 77189-940 77189-942 77189-944
2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid 97%
2-(4-Methyl-2-phenylpiperazin-1-yl)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₂
MW: 297.352 g/mol
MDL Number: MFCD08458719
CAS Number: 61338-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR