Methoxycarbonyl glycine 97%

Supplier: Ambeed
Warning

Synonyms: 2-((Methoxycarbonyl)amino)acetic acid

A127432-10G A127432-1G A127432-250MG A127432-25G A127432-5G
77161-664EA 131.84 USD
77161-664 77161-666 77161-668 77161-670 77161-672
Methoxycarbonyl glycine 97%
Methoxycarbonyl glycine

Precaución: for Research Use Only

Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00270561
CAS Number: 1670-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR