2-Fluoro-3-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A127075-100G A127075-1G A127075-25G A127075-5G
77189-926EA 471.74 USD
77189-926 77189-928 77189-930 77189-932
2-Fluoro-3-(trifluoromethyl)benzaldehyde 95%
2-Fluoro-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MW: 192.11 g/mol
MDL Number: MFCD00061246
CAS Number: 112641-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR