2',4'-Dichloro-5'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Dichloro-5-fluorophenyl)ethanone

A126646-100G A126646-10G A126646-25G A126646-500G
77189-902EA 31.93 USD
77189-902 77189-904 77189-906 77189-908
2',4'-Dichloro-5'-fluoroacetophenone 98%
2',4'-Dichloro-5'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
MDL Number: MFCD00077499
CAS Number: 704-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR