Methyl 3-iodo-1H-pyrrolo[3,2-b]pyridine-5-carboxylate 95%

Supplier: Ambeed
Warning

A126435-100MG A126435-1G A126435-250MG
77161-644EA 244.11 USD
77161-644 77161-646 77161-648
Methyl 3-iodo-1H-pyrrolo[3,2-b]pyridine-5-carboxylate 95%
Methyl 3-iodo-1H-pyrrolo[3,2-b]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇IN₂O₂
MW: 302.06 g/mol
MDL Number: MFCD12963008
CAS Number: 1190310-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR