(1,3-Dimethyl-1H-pyrazol-4-yl)methylamine 95%

Supplier: Ambeed
Danger

Synonyms: (1,3-Dimethyl-1H-pyrazol-4-yl)methanamine

A126338-1G A126338-250MG A126338-5G
77161-634EA 69.01 USD
77161-634 77161-636 77161-638
(1,3-Dimethyl-1H-pyrazol-4-yl)methylamine 95%
(1,3-Dimethyl-1H-pyrazol-4-yl)methylamine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃
MW: 125.17 g/mol
MDL Number: MFCD02055709
CAS Number: 400756-28-7
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR