5-Nitro-1H-indazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A126212-1G A126212-250MG A126212-5G
77161-618EA 110.21 USD
77161-618 77161-620 77161-622
5-Nitro-1H-indazole-3-carboxylic acid 95%
5-Nitro-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅N₃O₄
MW: 207.14 g/mol
MDL Number: MFCD05663993
CAS Number: 78155-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR