(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid hydrate(2:1) 98%

Supplier: Ambeed
Danger

Synonyms: Levofloxacin hydrate

A126121-10G A126121-25G A126121-500G A126121-5G A126121-100MG
77189-892EA 8.24 USD
77189-892 77189-894 77189-896 77189-898 77745-915
(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid hydrate(2:1) 98%
(S)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid hydrate(2:1)

Precaución: for Research Use Only

Formula: C₃₆H₄₂F₂N₆O₉
MW: 740.75 g/mol
MDL Number: MFCD07772024
CAS Number: 138199-71-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR