1-Chloro-3-fluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A125993-10G A125993-1G A125993-250MG
77161-612EA 103 USD
77161-612 77161-614 77161-616
1-Chloro-3-fluoro-5-nitrobenzene 98%
1-Chloro-3-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD11973918
CAS Number: 4815-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR