(2R,3R)-Methyl-2-amino-3-hydroxybutanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-allo-Thr-OMe·HCl, (2R,3R)-Methyl 2-amino-3-hydroxybutanoate hydrochloride
A125929-1G
A125929-250MG
A125929-5G
77161-606EA
129.78
USD
77161-606
77161-608
77161-610
(2R,3R)-Methyl-2-amino-3-hydroxybutanoate hydrochloride 97%
(2R,3R)-Methyl-2-amino-3-hydroxybutanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₃ MW: 169.605 g/mol |
MDL Number:
MFCD08458597 CAS Number: 60538-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...