3-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Isopropoxypyridine-3-boronic acid pinacol ester

A125822-250MG A125822-100MG A125822-1G
77161-598EA 75.19 USD
77161-598 77683-120 77683-121
3-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%
3-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: For research use only.

Formula: C₁₄H₂₂BNO₃
MW: 263.14 g/mol
MDL Number: MFCD13182246
CAS Number: 1171892-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR