7-Bromo-2,4,6-trichloroquinazoline 97%

Supplier: Ambeed
Warning

A125820-100MG A125820-1G A125820-250MG
77189-886EA 84.46 USD
77189-886 77189-888 77189-890
7-Bromo-2,4,6-trichloroquinazoline 97%
7-Bromo-2,4,6-trichloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₂BrCl₃N₂
MW: 312.38 g/mol
CAS Number: 1956389-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR