2-Fluoro-6-methoxybenzylamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Fluoro-6-methoxyphenyl)methanamine
A125617-10G
A125617-1G
A125617-25G
A125617-5G
77189-878EA
249.26
USD
77189-878
77189-880
77189-882
77189-884
2-Fluoro-6-methoxybenzylamine 98%
2-Fluoro-6-methoxybenzylamine
Precaución: for Research Use Only
Formula:
C₈H₁₀FNO MW: 155.1695 g/mol |
MDL Number:
MFCD00042292 CAS Number: 150517-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...