9-Mesityl-10-phenylacridin-10-ium tetrafluoroborate 98%

Supplier: Ambeed
Warning

A125582-100MG A125582-1G A125582-250MG A125582-5G
77189-870EA 12.36 USD
77189-870 77189-872 77189-874 77189-876
9-Mesityl-10-phenylacridin-10-ium tetrafluoroborate 98%
9-Mesityl-10-phenylacridin-10-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₈H₂₄BF₄N
MW: 461.30 g/mol
MDL Number: MFCD28411687
CAS Number: 1621019-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR