Benzyl (3-fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)carbamate 97%

Supplier: Ambeed
Danger

A125420-1G A125420-250MG
77189-862EA 63 USD
77189-862 77189-864
Benzyl (3-fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)carbamate 97%
Benzyl (3-fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₁H₁₇FN₆O₂
MW: 404.39 g/mol
MDL Number: MFCD22418002
CAS Number: 1220910-89-3
UN: 1325
ADR: 4.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR