5-Hydroxy-2-pentanone 92% (mixture of monomer and dimer)

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxypentan-2-one, 3-Acetyl-1-propanol

A125361-1000G A125361-100G A125361-10G A125361-25G A125361-500G
77161-580EA 109.18 USD
77161-580 77161-582 77161-584 77161-586 77161-588
5-Hydroxy-2-pentanone 92% (mixture of monomer and dimer)
5-Hydroxy-2-pentanone

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00002961
CAS Number: 1071-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR