(R)-tert-Butyl (2-oxopyrrolidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A125340-100MG A125340-1G A125340-250MG A125340-5G
77161-572EA 99.91 USD
77161-572 77161-574 77161-576 77161-578
(R)-tert-Butyl (2-oxopyrrolidin-3-yl)carbamate 95%
(R)-tert-Butyl (2-oxopyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
MW: 200.24 g/mol
MDL Number: MFCD11616145
CAS Number: 251938-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR