PF-3758309 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-N-(2-(Dimethylamino)-1-phenylethyl)-6,6-dimethyl-3-((2-methylthieno[3,2-d]pyrimidin-4-yl)amino)-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide
A125108-10MG
A125108-1MG
A125108-50MG
A125108-5MG
77189-846EA
61
USD
77189-846
77189-848
77189-850
77189-852
PF-3758309 98+%
PF-3758309
Precaución: for Research Use Only
Formula:
C₂₅H₃₀N₈OS MW: 490.62 g/mol |
MDL Number:
MFCD18633226 CAS Number: 898044-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...