4-Chloro-2,6-xylidine 98%

Supplier: Ambeed
Warning

A125101-1G A125101-250MG A125101-5G
77189-840EA 24 USD
77189-840 77189-842 77189-844
4-Chloro-2,6-xylidine 98%
4-Chloro-2,6-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MDL Number: MFCD01076576
CAS Number: 24596-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR