1,4-Dichloro-2,5-difluorobenzene 95+%

Supplier: Ambeed
Warning

A125097-1G A125097-5G A125097-25G
77161-552EA 39.14 USD
77161-552 77161-554 77611-066
1,4-Dichloro-2,5-difluorobenzene 95+%
1,4-Dichloro-2,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₂
MW: 182.98 g/mol
MDL Number: MFCD13193245
CAS Number: 400-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR