Methyl 4-amino-3,5-dichlorobenzoate 97%

Supplier: Ambeed
Warning

A125006-1G A125006-250MG A125006-25G A125006-5G
77161-536EA 12 USD
77161-536 77161-538 77161-540 77161-542
Methyl 4-amino-3,5-dichlorobenzoate 97%
Methyl 4-amino-3,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MDL Number: MFCD08436086
CAS Number: 41727-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR