Fatostatin hydrobromide 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Propylpyridin-4-yl)-4-(p-tolyl)thiazole hydrobromide, Fatostatin HBr

A124858-10MG A124858-1MG A124858-25MG A124858-50MG A124858-5MG
77161-518EA 56.65 USD
77161-518 77161-520 77161-522 77161-524 77161-526
Fatostatin hydrobromide 98%
Fatostatin hydrobromide

Precaución: for Research Use Only

Formula: C₁₈H₁₉BrN₂S
MW: 375.33 g/mol
MDL Number: MFCD02376000
CAS Number: 298197-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR