4-Bromo-1H-pyrazolo[4,3-c]pyridine 95%
Supplier: Ambeed
|
Warning
|
A124250-100MG
A124250-250MG
A124250-1G
77161-508EA
25.75
USD
77161-508
77161-510
77683-109
4-Bromo-1H-pyrazolo[4,3-c]pyridine 95%
4-Bromo-1H-pyrazolo[4,3-c]pyridine
Precaución: For research use only.
Formula:
C₆H₄BrN₃ |
MDL Number:
MFCD12401001 CAS Number: 1159829-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...