3-Chloro-4-(trifluoromethoxy)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-trifluoromethoxybenzaldehyde

A124172-100G A124172-10G A124172-1G A124172-25G A124172-5G
77189-808EA 469.68 USD
77189-808 77189-810 77189-812 77189-814 77189-816
3-Chloro-4-(trifluoromethoxy)benzaldehyde 98%
3-Chloro-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C8H4ClF3O2
MW: 224.5600 g/mol
MDL Number: MFCD01631561
CAS Number: 83279-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR