8-Oxa-2-azaspiro[4.5]decane oxalate(2:1) 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A124060-1G A124060-250MG A124060-100MG A124060-5G
77161-496EA 42 USD
77161-496 77161-498 77683-107 77891-883
8-Oxa-2-azaspiro[4.5]decane oxalate(2:1) 97%, Ambeed.Inc
8-Oxa-2-azaspiro[4.5]decane oxalate(2:1)

Precaución: For research use only.

Formula: C₁₈H₃₂N₂O₆
MDL Number: MFCD28054406
CAS Number: 1651840-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR