2-(Bromomethyl)-6-fluorobenzonitrile 97%

Supplier: Ambeed
Danger

A123826-100MG A123826-10G A123826-1G A123826-250MG A123826-25G
77189-754EA 17 USD
77189-754 77189-756 77189-758 77189-760 77189-762
2-(Bromomethyl)-6-fluorobenzonitrile 97%
2-(Bromomethyl)-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MDL Number: MFCD18394042
CAS Number: 1261686-95-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR