2-Phenoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenoxy-1-phenylethanone

A123693-1G A123693-250MG A123693-5G
77161-480EA 98.88 USD
77161-480 77161-482 77161-484
2-Phenoxyacetophenone 98%
2-Phenoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00156689
CAS Number: 721-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR