1-Bromo-4-chloro-2,3-dimethylbenzene 97%
Supplier: Ambeed
|
Warning
|
A123241-100MG
A123241-1G
A123241-250MG
77189-742EA
69.01
USD
77189-742
77189-744
77189-746
1-Bromo-4-chloro-2,3-dimethylbenzene 97%
1-Bromo-4-chloro-2,3-dimethylbenzene
Precaución: for Research Use Only
Formula:
C₈H₈BrCl MW: 219.50 g/mol |
MDL Number:
MFCD11846011 CAS Number: 425394-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...