(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride 97%

Supplier: Ambeed
Warning

A123141-10G A123141-1G A123141-250MG A123141-5G
77161-446EA 129.78 USD
77161-446 77161-448 77161-450 77161-452
(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride 97%
(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂NO
MW: 260.1600 g/mol
MDL Number: MFCD00053028
CAS Number: 55695-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR